Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000534.1 | XP_016856023.1 | 12 | 2341 | ||
XM_017000535.1 | XP_016856024.1 | 12 | 2301 | |||
XM_017000536.1 | XP_016856025.1 | 11 | 2149 | |||
GenBank mRNA | AK296788.1 | - | 11 | 1394 | ||
BC037166.2 | - | 12 | 2189 | 5090654 | ||
M59371.1 | - | 12 | 2184 |