Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPN1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012337.2 | NP_001012337.1 | 6 | 854 | ||
NM_001308313.1 | NP_001295242.1 | 7 | 900 | |||
XM_005247138.3 | XP_005247195.1 | 5 | 709 | |||
XM_006713513.3 | XP_006713576.1 | 8 | 996 | |||
GenBank mRNA | AF231410.1 | - | 6 | 813 | ||
BC015413.1 | - | 4 | 594 | 4394326 | ||
BC141849.1 | - | 7 | 847 | 8860347 |