Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152542.4 | NP_689755.3 | 4 | 1070 | ||
XM_006714111.3 | XP_006714174.1 | 4 | 1131 | |||
XM_017007803.1 | XP_016863292.1 | 4 | 718 | |||
XM_017007804.1 | XP_016863293.1 | 5 | 1258 | |||
XM_017007805.1 | XP_016863294.1 | 6 | 1258 | |||
XM_017007806.1 | XP_016863295.1 | 4 | 591 | |||
XM_017007807.1 | XP_016863296.1 | 3 | 309 | |||
GenBank mRNA | AF351614.1 | - | 5 | 991 | ||
AK054678.1 | - | 4 | 798 | |||
AK314417.1 | - | 4 | 795 | |||
AL834271.1 | - | 4 | 789 | |||
AX766363.1 | - | 3 | 585 | |||
AY157615.1 | - | 4 | 798 | |||
AY435431.1 | - | 4 | 975 | |||
BC020850.1 | - | 4 | 800 | |||
BC037552.1 | - | 4 | 834 | 5227317 |