Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129890.1 | NP_001123362.1 | 1 | 98 | ||
GenBank mRNA | AK097393.1 | - | 1 | 98 | ||
AK130865.1 | - | 1 | 72 |