Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126049.1 | NP_001119521.1 | 1 | 3787 | ||
GenBank mRNA | EU552092.1 | - | 1 | 3787 |