Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000247.2 | NP_000238.1 | 6 | 1357 | ||
NM_001177519.2 | NP_001170990.1 | 6 | 1361 | |||
NM_001289152.1 | NP_001276081.1 | 6 | 2225 | |||
NM_001289153.1 | NP_001276082.1 | 6 | 2205 | |||
NM_001289154.1 | NP_001276083.1 | 6 | 2033 | |||
GenBank mRNA | AK299135.1 | - | 6 | 1333 | ||
L14848.1 | - | 6 | 1330 |