Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM81B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152548.2 | NP_689761.2 | 3 | 320 | ||
XM_005271905.2 | XP_005271962.2 | 3 | 298 | |||
XM_005271906.4 | XP_005271963.2 | 4 | 343 | |||
XM_005271907.3 | XP_005271964.1 | 3 | 305 | |||
XM_011543207.1 | XP_011541509.1 | 3 | 315 | |||
XM_011543208.1 | XP_011541510.1 | 3 | 315 | |||
XM_017009118.1 | XP_016864607.1 | 4 | 340 | |||
GenBank mRNA | AK058062.1 | - | 3 | 303 | ||
AK310245.1 | - | 2 | 122 | |||
BC034772.1 | - | 3 | 320 | 5169904 | ||
BM560335.1 | - | 2 | 125 |