Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM81B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152548.2 | NP_689761.2 | 8 | 962 | ||
XM_005271905.2 | XP_005271962.2 | 7 | 821 | |||
XM_005271906.4 | XP_005271963.2 | 10 | 1126 | |||
XM_011543207.1 | XP_011541509.1 | 9 | 1046 | |||
XM_011543209.1 | XP_011541511.1 | 6 | 643 | |||
XM_011543210.2 | XP_011541512.1 | 5 | 676 | |||
XM_017009118.1 | XP_016864607.1 | 9 | 982 | |||
GenBank mRNA | AK058062.1 | - | 8 | 945 | ||
AK310245.1 | - | 8 | 905 | |||
BC034772.1 | - | 8 | 962 | 5169904 |