Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102426.1 | NP_001095896.1 | 2 | 352 | ||
XM_005263862.3 | XP_005263919.1 | 2 | 596 | |||
XM_005263863.4 | XP_005263920.1 | 2 | 383 | |||
XM_005263864.3 | XP_005263921.1 | 2 | 593 | |||
XM_011510507.1 | XP_011508809.1 | 4 | 438 | |||
XM_011510508.2 | XP_011508810.1 | 2 | 352 | |||
XM_017003198.1 | XP_016858687.1 | 2 | 608 | |||
XM_017003199.1 | XP_016858688.1 | 2 | 606 | |||
XM_017003200.1 | XP_016858689.1 | 2 | 626 | |||
XM_017003201.1 | XP_016858690.1 | 2 | 617 | |||
XM_017003202.1 | XP_016858691.1 | 2 | 652 | |||
GenBank mRNA | AB449889.1 | - | 2 | 221 | ||
AK300533.1 | - | 2 | 352 | |||
DB201187.1 | - | 2 | 350 | |||
DC383710.1 | - | 2 | 352 | |||
DR001338.1 | - | 1 | 48 |