Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126522.1 | 1 | 762 | |||
GenBank mRNA | BC014011.2 | - | 1 | 697 | ||
BC029796.1 | - | 1 | 745 | 5183143 |