Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA5B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039500.2 | NP_001034589.2 | 22 | 3994 | ||
XM_006710360.3 | XP_006710423.2 | 19 | 3269 | |||
XM_011540683.2 | XP_011538985.1 | 22 | 3956 | |||
XM_011540684.2 | XP_011538986.1 | 22 | 4081 | |||
XM_011540685.2 | XP_011538987.1 | 22 | 4093 | |||
XM_011540688.2 | XP_011538990.1 | 21 | 3881 | |||
XM_011540690.2 | XP_011538992.1 | 21 | 3779 | |||
XM_011540691.2 | XP_011538993.1 | 21 | 3770 | |||
XM_011540692.2 | XP_011538994.1 | 22 | 3950 | |||
XM_011540697.2 | XP_011538999.1 | 16 | 3327 | |||
XM_017000305.1 | XP_016855794.1 | 16 | 3073 | |||
XM_017000306.1 | XP_016855795.1 | 17 | 3234 |