Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173615.4 | NP_775886.3 | 15 | 1523 | ||
XM_011545742.2 | XP_011544044.1 | 17 | 1670 | |||
XM_011545744.2 | XP_011544046.1 | 2 | 299 | |||
XM_011545745.1 | XP_011544047.1 | 1 | 45 | |||
XM_011545746.2 | XP_011544048.1 | 2 | 299 | |||
XM_017022947.1 | XP_016878436.1 | 18 | 1743 | |||
XM_017022948.1 | XP_016878437.1 | 1 | 871 | |||
XM_017022949.1 | XP_016878438.1 | 1 | 871 | |||
GenBank mRNA | AK098260.1 | - | 13 | 1480 | ||
AK128606.1 | - | 4 | 391 | |||
AK307544.1 | - | 15 | 1519 | |||
BC109294.1 | - | 3 | 251 | |||
BC109295.1 | - | 3 | 251 |