Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100915.1 | NP_001094385.1 | 9, 10 | 1437 | ||
GenBank mRNA | AK097481.1 | - | 9, 10 | 1440 | ||
AK302567.1 | - | 8, 9 | 2655 | |||
AK302748.1 | - | 9, 10 | 1421 | |||
BC070103.1 | - | 9, 10 | 3218 | |||
BC094790.1 | - | 7, 8 | 3384 | |||
BC117335.1 | - | 9, 10 | 1437 | 40125886 |