Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207305.4 | NP_997188.2 | 1 | 308 | ||
GenBank mRNA | AY344640.1 | - | 1 | 96 | ||
BC089432.1 | - | 1 | 311 | 30555309 | ||
BC136570.1 | - | 1 | 43 | 9020560 | ||
BC136571.1 | - | 1 | 43 | 9020561 | ||
CF139066.1 | - | 1 | 327 |