Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099784.2 | NP_001093254.2 | 2 | 333 | ||
NM_001282351.1 | NP_001269280.1 | 2 | 199 | |||
GenBank mRNA | AK127701.1 | - | 2 | 183 | ||
AK302669.1 | - | 2 | 335 | |||
CX781342.1 | - | 2 | 179 | |||
DA244744.1 | - | 2 | 238 | |||
DC325425.1 | - | 1 | 115 | |||
DC401737.1 | - | 2 | 336 |