Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323916.1 | NP_001310845.1 | 17 | 4698 | ||
NM_020840.2 | NP_065891.1 | 17 | 4693 | |||
XM_005263156.1 | XP_005263213.1 | 18 | 4783 | |||
XM_005263158.1 | XP_005263215.1 | 18 | 4583 | |||
XM_011532154.2 | XP_011530456.1 | 18 | 5563 | |||
XM_017008487.1 | XP_016863976.1 | 19 | 4732 | |||
GenBank mRNA | AB040883.1 | - | 17 | 4550 | ||
AK022968.1 | - | 5 | 2490 | |||
BC047069.1 | - | 1 | 533 |