Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039763.3 | NP_001034852.3 | 2 | 146 | ||
XM_006714670.3 | XP_006714733.1 | 4 | 522 | |||
XM_011543552.2 | XP_011541854.1 | 5 | 873 | |||
XM_011543553.2 | XP_011541855.1 | 2 | 956 | |||
XM_011543557.2 | XP_011541859.1 | 5 | 874 | |||
XM_011543559.2 | XP_011541861.1 | 5 | 873 | |||
XM_011543561.2 | XP_011541863.1 | 5 | 873 | |||
XM_011543563.2 | XP_011541865.1 | 5 | 872 | |||
XM_011543564.2 | XP_011541866.1 | 5 | 871 | |||
XM_011543565.2 | XP_011541867.1 | 5 | 871 | |||
XM_011543567.2 | XP_011541869.1 | 5 | 871 | |||
XM_017009704.1 | XP_016865193.1 | 3 | 1102 | |||
XM_017009705.1 | XP_016865194.1 | 5 | 874 | |||
XM_017009706.1 | XP_016865195.1 | 5 | 873 | |||
XM_017009707.1 | XP_016865196.1 | 4 | 523 | |||
GenBank mRNA | AK296542.1 | - | 2 | 62 | ||
BX648527.1 | - | 2 | 69 | |||
DC399208.1 | - | 2 | 77 |