Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098612.1 | NP_001092082.1 | 7 | 1604 | ||
XM_017026113.1 | XP_016881602.1 | 6 | 1292 | |||
GenBank mRNA | AY854038.1 | - | 7 | 1525 |