Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167961.2 | NP_001161433.1 | 4 | 998 | ||
NM_153033.4 | NP_694578.1 | 4 | 998 | |||
GenBank mRNA | AK055201.1 | - | 2 | 337 | ||
AK056631.1 | - | 4 | 921 | |||
BC042482.1 | - | 4 | 936 | 5165722 |