Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258211.1 | NP_001245140.1 | 5 | 808 | ||
NM_172366.3 | NP_758954.1 | 6 | 844 | |||
GenBank mRNA | AF453435.1 | - | 6 | 753 | ||
AL560247.3 | - | 5 | 768 | |||
BC074986.2 | - | 6 | 743 | 30915303 | ||
BC102026.1 | - | 6 | 763 | 40031445 | ||
BC102027.1 | - | 5 | 727 | 40031444 | ||
BC102028.1 | - | 5 | 727 | 40031440 |
Gene Symbol: ZNF395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098243.1 | - | 6 | 803 |