Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ERICH1 SIRNA Assays ›
Gene Symbol:
ERICH1
Gene Name:
glutamate rich 1
Gene Aliases:
HSPC319
Chromosome Location:
Chr.8: 614737 - 731239 on Build GRCh38
UniGene ID:
Hs.655310
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s45985
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ERICH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303100.1
NP_001290029.1
5
1269
NM_207332.2
NP_997215.1
5
1269
XM_006716234.3
XP_006716297.2
5
1258
XM_011534731.2
XP_011533033.1
6
1441
XM_011534735.2
XP_011533037.1
5
1258
XM_011534739.2
XP_011533041.1
5
1248
XM_017013114.1
XP_016868603.1
6
1456
XM_017013115.1
XP_016868604.1
6
1456
XM_017013116.1
XP_016868605.1
6
1456
XM_017013117.1
XP_016868606.1
6
1441
XM_017013118.1
XP_016868607.1
6
1456
XM_017013119.1
XP_016868608.1
6
1456
XM_017013120.1
XP_016868609.1
6
1456
XM_017013121.1
XP_016868610.1
6
1456
XM_017013122.1
XP_016868611.1
6
1456
XM_017013123.1
XP_016868612.1
6
1456
XM_017013124.1
XP_016868613.1
6
1456
XM_017013125.1
XP_016868614.1
6
1456
XM_017013126.1
XP_016868615.1
6
1456
XM_017013127.1
XP_016868616.1
6
1456
XM_017013132.1
XP_016868621.1
5
1258
XM_017013134.1
XP_016868623.1
5
1248
GenBank mRNA
AK125470.1
-
2
170
AK290264.1
-
5
1253
AK297476.1
-
5
1239
AL833324.1
-
2
497
BC016017.1
-
336
BC046243.1
-
5
1256
5207311
BX647093.1
-
2
1678
CN353744.1
-
1
613
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0