Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM120AOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322224.1 | NP_001309153.1 | 3 | 657 | ||
NM_198841.2 | NP_942138.2 | 3 | 1586 | |||
NR_136229.1 | 3 | 993 | ||||
NR_136230.1 | 4 | 1114 | ||||
NR_136231.1 | 4 | 1707 | ||||
NR_136232.1 | 4 | 921 | ||||
NR_136233.1 | 3 | 744 | ||||
NR_136234.1 | 4 | 778 | ||||
NR_136235.1 | 3 | 800 | ||||
NR_136236.1 | 3 | 1003 | ||||
NR_136237.1 | 4 | 1124 | ||||
NR_136238.1 | 4 | 865 | ||||
GenBank mRNA | AK056096.1 | - | 3 | 1586 | ||
AK309269.1 | - | 4 | 676 | |||
BC063650.1 | - | 3 | 550 | |||
BC080634.1 | - | 4 | 648 | |||
BC107135.1 | - | 4 | 779 | |||
BC107136.1 | - | 3 | 658 | |||
BC113074.1 | - | 3 | 658 |