Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102576.2 | NP_001096046.2 | 3 | 266 | ||
NM_153478.2 | NP_705611.2 | 4 | 482 | |||
GenBank mRNA | AF195880.1 | - | 4 | 480 | ||
AF268418.1 | - | 3 | 236 | |||
AF268419.1 | - | 4 | 433 | |||
BC059947.1 | - | 3 | 236 | 5397842 | ||
BF570999.1 | - | 3 | 265 | |||
BM810082.1 | - | 3 | 248 |