Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134658.1 | NP_001128130.1 | 3 | 655 | ||
NM_153226.2 | NP_694958.1 | 3 | 652 | |||
XM_011539371.2 | XP_011537673.1 | 3 | 620 | |||
GenBank mRNA | AK091309.1 | - | 3 | 652 | ||
BC041432.1 | - | 2 | 384 | |||
BC104814.1 | - | 3 | 626 | 8143817 | ||
BC143283.1 | - | 3 | 575 | 9051786 | ||
BC143284.1 | - | 3 | 626 | 9051787 |