Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178498.3 | NP_848593.2 | 5 | 915 | ||
XM_006718155.3 | XP_006718218.1 | 5 | 818 | |||
XM_006718156.3 | XP_006718219.1 | 5 | 1154 | |||
XM_011519920.2 | XP_011518222.1 | 6 | 395 | |||
XM_011519921.2 | XP_011518223.1 | 5 | 1155 | |||
XM_017017244.1 | XP_016872733.1 | 6 | 841 | |||
GenBank mRNA | AY299482.1 | - | 5 | 624 | ||
BC041454.1 | - | 5 | 384 | |||
BC049207.1 | - | 5 | 887 | 5188979 |