Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145913.3 | NP_666018.3 | 5 | 993 | ||
XM_017018910.1 | XP_016874399.1 | 5 | 1029 | |||
GenBank mRNA | AF536216.1 | - | 5 | 966 | ||
AK075263.1 | - | 5 | 651 | |||
AY081220.1 | - | 5 | 967 | |||
BC110492.2 | - | 5 | 628 | 40007567 |