Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144033.1 | NP_001137505.1 | 6 | 707 | ||
NM_145286.2 | NP_660329.1 | 5 | 623 | |||
XM_017020400.1 | XP_016875889.1 | 3 | 331 | |||
GenBank mRNA | AK304159.1 | - | 6 | 707 | ||
AK313632.1 | - | 5 | 595 | |||
BC025760.1 | - | 5 | 610 | 5204252 |