Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145545.1 | NP_001139017.1 | 1 | 28 | ||
GenBank mRNA | AF536240.1 | - | 1 | 219 | ||
BC136370.1 | - | 1 | 211 | 9020357 | ||
BC136382.1 | - | 1 | 211 | 9020369 |