Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153228.2 | NP_694960.2 | 13 | 1538 | ||
XM_006721728.3 | XP_006721791.1 | 12 | 1761 | |||
XM_011524428.2 | XP_011522730.1 | 13 | 1538 | |||
XM_011524429.2 | XP_011522731.1 | 13 | 1695 | |||
XM_011524430.2 | XP_011522732.1 | 14 | 1661 | |||
XM_011524431.2 | XP_011522733.1 | 13 | 1593 | |||
XM_011524433.2 | XP_011522735.1 | 7 | 951 | |||
XM_011524434.2 | XP_011522736.1 | 4 | 398 | |||
XM_017024263.1 | XP_016879752.1 | 15 | 1755 | |||
XM_017024264.1 | XP_016879753.1 | 15 | 2740 | |||
XM_017024265.1 | XP_016879754.1 | 15 | 1949 | |||
XM_017024266.1 | XP_016879755.1 | 13 | 1710 | |||
XM_017024267.1 | XP_016879756.1 | 12 | 1562 | |||
XM_017024268.1 | XP_016879757.1 | 14 | 2646 | |||
XM_017024269.1 | XP_016879758.1 | 12 | 1835 | |||
XM_017024270.1 | XP_016879759.1 | 15 | 1753 | |||
GenBank mRNA | AK095654.1 | - | 13 | 1538 |