Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207333.2 | NP_997216.2 | 4 | 1123 | ||
XM_006723059.3 | XP_006723122.1 | 5 | 1169 | |||
XM_017026393.1 | XP_016881882.1 | 6 | 1414 | |||
XM_017026394.1 | XP_016881883.1 | 6 | 1339 | |||
XM_017026395.1 | XP_016881884.1 | 6 | 1487 | |||
XM_017026396.1 | XP_016881885.1 | 5 | 1256 | |||
XM_017026397.1 | XP_016881886.1 | 5 | 1261 | |||
XM_017026398.1 | XP_016881887.1 | 5 | 1140 | |||
XM_017026399.1 | XP_016881888.1 | 4 | 1133 | |||
XM_017026400.1 | XP_016881889.1 | 4 | 1053 | |||
GenBank mRNA | AK129992.1 | - | 1 | 660 | ||
AL831902.1 | - | 1 | 321 | |||
BC105738.1 | - | 8 | 1435 | |||
BC131555.1 | - | 3 | 978 | 40108031 | ||
CR933622.1 | - | 2 | 909 |