Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139284.2 | NP_644813.1 | 3 | 862 | ||
XM_011526595.1 | XP_011524897.1 | 3 | 216 | |||
XM_017026428.1 | XP_016881917.1 | 3 | 191 | |||
XM_017026429.1 | XP_016881918.1 | 3 | 191 | |||
GenBank mRNA | AF467954.1 | - | 3 | 554 | ||
AJ487519.1 | - | 3 | 787 | |||
AJ487959.1 | - | 3 | 295 | |||
AY358121.1 | - | 3 | 457 | |||
BC029943.1 | - | 4 | 327 | |||
BC087848.1 | - | 3 | 439 | |||
BC136694.1 | - | 3 | 337 | 9020684 | ||
BC136697.1 | - | 3 | 337 | 9020687 |