Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039876.2 | NP_001034965.1 | 6 | 1054 | ||
NM_001297735.2 | NP_001284664.1 | 4 | 715 | |||
GenBank mRNA | AK093764.1 | - | 5 | 2013 | ||
BC052573.1 | - | 4 | 705 | 6568945 |