Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199085.1 | NP_001186014.1 | 10 | 2065 | ||
NM_001199089.1 | NP_001186018.1 | 10 | 1778 | |||
NM_001199091.1 | NP_001186020.1 | 10 | 1906 | |||
NM_001199092.1 | NP_001186021.1 | 7 | 750 | |||
NM_173533.3 | NP_775804.2 | 10 | 2065 | |||
XM_005244934.1 | XP_005244991.1 | 10 | 1850 | |||
XM_005244935.4 | XP_005244992.1 | 10 | 1755 | |||
XM_017000473.1 | XP_016855962.1 | 10 | 1771 | |||
GenBank mRNA | AK092142.1 | - | 7 | 763 | ||
AK126889.1 | - | 10 | 1906 | |||
AK127370.1 | - | 10 | 1797 | |||
BC065707.1 | - | 7 | 874 | |||
BC089387.1 | - | 8 | 967 | |||
BC130532.1 | - | 10 | 1742 | 40146563 | ||
BC144058.1 | - | 10 | 1797 | 9052578 |