Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199085.1 | NP_001186014.1 | 9 | 1996 | ||
NM_001199089.1 | NP_001186018.1 | 9 | 1709 | |||
NM_001199091.1 | NP_001186020.1 | 9 | 1837 | |||
NM_001199092.1 | NP_001186021.1 | 6 | 681 | |||
NM_173533.3 | NP_775804.2 | 9 | 1996 | |||
XM_005244934.1 | XP_005244991.1 | 9 | 1781 | |||
XM_005244935.4 | XP_005244992.1 | 9 | 1686 | |||
XM_017000473.1 | XP_016855962.1 | 9 | 1702 | |||
GenBank mRNA | AK092142.1 | - | 6 | 694 | ||
AK126889.1 | - | 9 | 1837 | |||
AK127370.1 | - | 9 | 1728 | |||
BC065707.1 | - | 6 | 805 | |||
BC089387.1 | - | 7 | 898 | |||
BC130532.1 | - | 9 | 1673 | 40146563 | ||
BC144058.1 | - | 9 | 1728 | 9052578 |