Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALML6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138705.2 | NP_619650.2 | 4, 5 | 857 | ||
XM_005244729.3 | XP_005244786.2 | 4, 5 | 1999 | |||
XM_011540841.2 | XP_011539143.1 | 3, 4 | 1882 | |||
GenBank mRNA | AF490905.2 | - | 4, 5 | 857 | ||
AY566229.1 | - | 4, 5 | 857 |