Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113523.2 | NP_001106995.1 | 12 | 1956 | ||
NM_001308320.1 | NP_001295249.1 | 9 | 1371 | |||
NM_001308321.1 | NP_001295250.1 | 7 | 1076 | |||
NM_152615.2 | NP_689828.1 | 8 | 1283 | |||
XM_005247159.3 | XP_005247216.1 | 11 | 3421 | |||
XM_005247160.3 | XP_005247217.1 | 11 | 3400 | |||
XM_011512475.2 | XP_011510777.1 | 11 | 3512 | |||
XM_011512476.2 | XP_011510778.1 | 11 | 3481 | |||
XM_011512477.2 | XP_011510779.1 | 10 | 3250 | |||
XM_011512478.2 | XP_011510780.1 | 12 | 1739 | |||
XM_011512479.2 | XP_011510781.1 | 10 | 1214 | |||
XM_017005791.1 | XP_016861280.1 | 10 | 3193 | |||
XM_017005792.1 | XP_016861281.1 | 9 | 2381 | |||
GenBank mRNA | AK097515.1 | - | 14 | 1901 | ||
AK097916.1 | - | 8 | 1220 | |||
BC101701.1 | - | 8 | 1283 | 8069207 | ||
BC101703.1 | - | 8 | 1283 | 8069209 | ||
BC143573.1 | - | 7 | 1076 | 9052084 | ||
BX647656.1 | - | 9 | 1347 | |||
DQ063586.1 | - | 12 | 1875 |