Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRICKLE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198859.3 | NP_942559.1 | 6, 7 | 1374 | ||
XM_011533432.2 | XP_011531734.1 | 7, 8 | 1168 | |||
XM_011533433.2 | XP_011531735.1 | 7, 8 | 1168 | |||
XM_011533434.2 | XP_011531736.1 | 7, 8 | 993 | |||
XM_011533435.2 | XP_011531737.1 | 7, 8 | 993 | |||
XM_011533436.2 | XP_011531738.1 | 6, 7 | 1226 | |||
XM_011533437.2 | XP_011531739.1 | 6, 7 | 10296 | |||
XM_011533438.2 | XP_011531740.1 | 3, 4 | 1118 | |||
XM_011533440.2 | XP_011531742.1 | 7, 8 | 1168 | |||
XM_017005798.1 | XP_016861287.1 | 6, 7 | 1098 | |||
XM_017005799.1 | XP_016861288.1 | 5, 6 | 2847 | |||
GenBank mRNA | AL833539.1 | - | 6, 7 | 1375 | ||
BC119002.1 | - | 6, 7 | 1355 | 40082599 | ||
BX647797.1 | - | 1, 2 | 70 |