Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRICKLE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198859.3 | NP_942559.1 | 3 | 750 | ||
XM_011533432.2 | XP_011531734.1 | 4 | 544 | |||
XM_011533433.2 | XP_011531735.1 | 4 | 544 | |||
XM_011533434.2 | XP_011531736.1 | 4 | 369 | |||
XM_011533435.2 | XP_011531737.1 | 4 | 369 | |||
XM_011533436.2 | XP_011531738.1 | 3 | 602 | |||
XM_011533437.2 | XP_011531739.1 | 3 | 9672 | |||
XM_011533440.2 | XP_011531742.1 | 4 | 544 | |||
XM_017005798.1 | XP_016861287.1 | 3 | 474 | |||
XM_017005799.1 | XP_016861288.1 | 2 | 2223 | |||
GenBank mRNA | AL833539.1 | - | 3 | 751 | ||
BC119002.1 | - | 3 | 731 | 40082599 |