Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040260.3 | NP_001035350.2 | 7 | 1970 | ||
NM_001040261.4 | NP_001035351.4 | 8 | 2021 | |||
NR_036614.1 | 7 | 1967 | ||||
XM_005262785.2 | XP_005262842.1 | 8 | 1847 | |||
XM_005262786.2 | XP_005262843.1 | 7 | 1796 | |||
XM_017007832.1 | XP_016863321.1 | 8 | 1850 | |||
XM_017007833.1 | XP_016863322.1 | 8 | 1850 | |||
GenBank mRNA | AB209181.1 | - | 7 | 1970 | ||
AL834498.1 | - | 8 | 1068 | |||
BC032726.1 | - | 7 | 1260 | |||
DA201927.1 | - | 5 | 417 |