Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122769.2 | NP_001116241.1 | 3, 4 | 1178 | ||
NM_181714.3 | NP_859065.2 | 4, 5 | 1333 | |||
XM_005248665.4 | XP_005248722.1 | 3, 4 | 1192 | |||
XM_011535504.1 | XP_011533806.1 | 4, 5 | 1245 | |||
GenBank mRNA | BC050327.1 | - | 4, 5 | 1333 | 4823983 | |
BX648161.1 | - | 3, 4 | 1159 |