Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXLNB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153235.3 | NP_694967.3 | 8 | 1278 | ||
XM_005266836.1 | XP_005266893.1 | 9 | 1645 | |||
XM_011535505.2 | XP_011533807.1 | 8 | 1382 | |||
XM_011535506.2 | XP_011533808.1 | 7 | 828 | |||
XM_017010322.1 | XP_016865811.1 | 9 | 1508 | |||
GenBank mRNA | AB085905.1 | - | 8 | 1278 | ||
AK314725.1 | - | 8 | 1226 | |||
AL831988.1 | - | 8 | 1216 | |||
AL832322.1 | - | 5 | 576 | |||
AL834248.2 | - | 8 | 1206 | |||
BC115383.1 | - | 8 | 1233 | 40038091 | ||
BC115384.2 | - | 8 | 1233 | 40038098 | ||
BX647477.1 | - | 8 | 1226 | |||
BX647633.1 | - | 8 | 1204 |