Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DACT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286350.1 | NP_001273279.1 | 3 | 1675 | ||
NM_214462.4 | NP_999627.2 | 4 | 2198 | |||
NR_104425.1 | 2 | 1207 | ||||
XM_011535507.2 | XP_011533809.1 | 4 | 1960 | |||
GenBank mRNA | AF318336.1 | - | 4 | 2166 | ||
BC111764.1 | - | 2 | 1163 | |||
BC111790.1 | - | 3 | 1631 |