Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173538.2 | NP_775809.1 | 7 | 866 | ||
XM_017013149.1 | XP_016868638.1 | 7 | 872 | |||
GenBank mRNA | AK093121.1 | - | 7 | 869 | ||
BC117326.1 | - | 7 | 866 | 40125877 | ||
BC117328.1 | - | 7 | 866 | 40125879 |