Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172811.1 | NP_001166282.1 | 9, 10 | 1129 | ||
NM_001172813.1 | NP_001166284.1 | 10, 11 | 1374 | |||
NM_001172814.1 | NP_001166285.1 | 8, 9 | 1216 | |||
NM_001172815.2 | NP_001166286.1 | 10, 11 | 1243 | |||
NM_173851.2 | NP_776250.2 | 7, 8 | 1186 | |||
GenBank mRNA | AK298294.1 | - | 11, 12 | 1377 | ||
AL713790.1 | - | 10, 11 | 1215 | |||
AY117411.1 | - | 7, 8 | 956 | |||
BC126446.1 | - | 8, 9 | 1055 | |||
BC143953.1 | - | 8, 9 | 1056 | |||
EF560713.1 | - | 9, 10 | 1129 |