Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010862.2 | NP_001010862.2 | 2 | 855 | ||
XM_006724577.3 | XP_006724640.1 | 2 | 916 | |||
XM_006724578.3 | XP_006724641.1 | 2 | 823 | |||
XM_017029315.1 | XP_016884804.1 | 2 | 1258 | |||
GenBank mRNA | AK127485.1 | - | 1 | 305 | ||
AK304047.1 | - | 2 | 792 | |||
AL832091.1 | - | 2 | 512 | |||
BC146899.1 | - | 2 | 576 | 9056827 | ||
BC146906.1 | - | 2 | 576 | 9056834 |