Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128933.2 | NP_001122405.1 | 2, 3 | 464 | ||
NM_001128934.2 | NP_001122406.1 | 2, 3 | 464 | |||
NM_001286754.1 | NP_001273683.1 | 2, 3 | 416 | |||
NM_133477.2 | NP_597734.2 | 2, 3 | 464 | |||
GenBank mRNA | AK307711.1 | - | 2, 3 | 460 | ||
AL832031.1 | - | 2, 3 | 450 | |||
AL832363.1 | - | 2, 3 | 358 | |||
AL833294.1 | - | 2, 3 | 450 | |||
AL833547.1 | - | 2, 3 | 464 | |||
BC136256.1 | - | 2, 3 | 339 | 9020243 | ||
BC150629.1 | - | 2, 3 | 463 | 9056999 | ||
BX648090.1 | - | 1, 2 | 122 | |||
CR936673.1 | - | 2, 3 | 456 | |||
EU481975.1 | - | 2, 3 | 268 | |||
KF147165.1 | - | 2, 3 | 416 |