Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030059.2 | NP_001025230.1 | 2 | 484 | ||
NM_001318166.1 | NP_001305095.1 | 2 | 484 | |||
NM_001318167.1 | NP_001305096.1 | 2 | 484 | |||
NM_001318168.1 | NP_001305097.1 | 2 | 484 | |||
NM_001318169.1 | NP_001305098.1 | 2 | 484 | |||
NR_134516.1 | 3 | 276 | ||||
XM_005269592.2 | XP_005269649.1 | 2 | 484 | |||
XM_006717685.3 | XP_006717748.1 | 2 | 153 | |||
XM_006717686.3 | XP_006717749.1 | 2 | 484 | |||
XM_011539444.2 | XP_011537746.1 | 3 | 211 | |||
XM_011539445.2 | XP_011537747.1 | 2 | 484 | |||
XM_017015820.1 | XP_016871309.1 | 2 | 156 | |||
XM_017015823.1 | XP_016871312.1 | 2 | 484 | |||
XM_017015824.1 | XP_016871313.1 | 2 | 484 | |||
GenBank mRNA | AK098668.1 | - | 2 | 102 | ||
AK296273.1 | - | 2 | 191 | |||
AK310178.1 | - | 2 | 135 | |||
BC101267.2 | - | 3 | 282 | |||
BC101268.2 | - | 3 | 178 | 40024099 | ||
BC101269.1 | - | 3 | 159 | |||
BC122535.1 | - | 3 | 281 | |||
BC132787.1 | - | 2 | 307 | 40146809 | ||
BI550027.1 | - | 2 | 216 | |||
DB473853.2 | - | 2 | 191 | |||
DB495310.2 | - | 2 | 461 | |||
DR000933.1 | - | 2 | 415 |