Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113402.1 | NP_001106873.1 | 4 | 623 | ||
NM_001278411.1 | NP_001265340.1 | 4 | 603 | |||
NM_001278412.1 | NP_001265341.1 | 6 | 832 | |||
NR_103522.1 | 5 | 699 | ||||
NR_103523.1 | 3 | 490 | ||||
XM_006719054.3 | XP_006719117.1 | 3 | 3916 | |||
XM_011520604.2 | XP_011518906.1 | 3 | 389 | |||
XM_017018963.1 | XP_016874452.1 | 6 | 672 | |||
XM_017018964.1 | XP_016874453.1 | 4 | 605 | |||
XM_017018965.1 | XP_016874454.1 | 5 | 697 | |||
GenBank mRNA | AK123033.1 | - | 4 | 470 | ||
AK302112.1 | - | 6 | 679 | |||
AK311214.1 | - | 3 | 416 | |||
AK311249.1 | - | 5 | 546 | |||
AL833434.1 | - | 4 | 473 | |||
BC037897.2 | - | 4 | 527 | 5275850 | ||
BC045831.1 | - | 4 | 567 | 5265088 | ||
BC067906.1 | - | 4 | 478 | 5314530 | ||
BI464068.1 | - | 4 | 556 |