Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173542.3 | NP_775813.2 | 7, 8 | 1147 | ||
XM_011538023.2 | XP_011536325.1 | 6, 7 | 930 | |||
XM_017018977.1 | XP_016874466.1 | 6, 7 | 1043 | |||
GenBank mRNA | BC030618.2 | - | 7, 8 | 1140 | 4822591 |