Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153486.3 | NP_705690.2 | 9, 10 | 1304 | ||
NM_194436.2 | NP_919417.1 | 9, 10 | 1235 | |||
GenBank mRNA | AY092767.1 | - | 9, 10 | 1240 | ||
BC040279.1 | - | 9, 10 | 1269 | 5162826 | ||
BC047902.1 | - | 9, 10 | 1299 | 5762691 |